missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Digital Dial Thermometer

Catalog No. S54110F
Change view
Click to view available options
Scale:
Centigrade, Fahrenheit Scale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale
S54110F Centigrade, Fahrenheit Scale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S54110F Supplier Fisherbrand™ Supplier No. 4042 EMD
Only null left

Easy to read digital dial and mercury free. Traceable™ Digital Dial Thermometers are ideal for cuvets, soil, incubators and test tubes. Wide temperature span combines the ranges of more than 90 different mercury thermometers.

  • LCD display 20 times more readable than eye-straining pointers or mercury levels
  • Readings updated every 10 seconds
  • Operate continuously for 1.5 years on a single replaceable battery
  • Resolution, Accuracy: 0.10, ±1°C Stem length: 4.8 in. (12 cm)
  • Stem diameter: 0.12 in. (3 mm)
  • Dial diameter: 1.5 in. (38 mm)
  • Weight: 1.01 oz. (31 g)
  • Range: -58°F to +572°F and -50°C to +300°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.19 in.
Diameter (Metric) Probe 4.83 mm
For Use With (Application) Soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum Hydrocarbons
Includes Protective Case, Silver Oxide Battery
Length (English) Probe 4.8 in.
Length (Metric) 18 cm
Length (Metric) Probe 12 cm
Material Plastic
Resolution 0.1°
Weight (English) 1 oz.
Weight (Metric) 31 g
Product Type Thermometer
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Battery Type Silver Oxide
Accuracy ±1°C (±1°F)
Display Digital LCD
Width (Metric) 5 cm
Scale Centigrade, Fahrenheit Scale
Show More Show Less