missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ 100-Hour Four-Channel Timer

Catalog No. 15079697
Change view
Click to view available options
Time Range:
99 hr. 59 min. and 59 sec. on 4 channels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Time Range
15079697 99 hr. 59 min. and 59 sec. on 4 channels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15079697 Supplier Fisherbrand™ Supplier No. 9001274

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand™ Traceable™ 100-Hour Four-Channel Timer is a lab workhorse provides 100 hour timing on four channels.

  • Alarm sounds for 60 seconds and may be silenced manually
  • Includes count/down feature and time-of-day clock

Specifications

Product Type Timer
Time Range 99 hr. 59 min. and 59 sec. on 4 channels
Alarms Audible
Accuracy 0.01%
Display Digital
Description USB Interface
No. of Channels 4
Battery Type AAA
Length (Metric) 10.16 cm
Weight (English) 1.5 oz.
Weight (Metric) 0.04 kg
Includes Battery, Flip-Open Stand, Traceable™ Certificate
Show More Show Less