Learn More
TR40 Concentrate
Catalog No. K1031
Encompass
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
K1031 1 L
1 options
Catalog No. K1031 Supplier Thermo Fisher Scientific Supplier No. LSK1031
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
TR40 Concentrate
TR40 Concentrate is the suppression dye component of Solution C in the Loading Kits for CCF2-AM and CCF4-AM.
Specifications
For Use With (Equipment) CCF2-AM and CCF4-AM
Product Line GeneBLAzer
Quantity 1 L
For Research Use Only. Not for use in diagnostic procedures.