missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TR-FRET Coregulator Buffer B

Catalog No. PV4418
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4418 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4418 Supplier Thermo Scientific™ Supplier No. PV4418
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

Specifications

For Use With (Equipment) LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays
Solubility Water soluble
For Use With (Application) Pharma and biopharma, target and lead identification and validation, drug discovery and development, nuclear receptor biology, biochemical nuclear receptor assays, nuclear receptor coregulator interaction
Description TR-FRET Coregulator Buffer B, 25mL, Liquid, Store at 20 to 30°C, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation
Content And Storage TR-FRET Coregulator Buffer B may be shipped frozen or at room temperature.Store at room temperature (20°C to 30°C).
Quantity 25 mL
Form Liquid
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
I have no assay window in my TR-FRET assay. What is the reason?

The single most common reason that a TR-FRET assay fails is that an incorrect choice of emission filters was made. Unlike other fluorescent assays, the filters used in a TR-FRET assay must be exactly those that are recommended for your instrument as the emission filter choice can make or break the assay. The excitation filter has more of an impact on the assay window. Please refer to our instrument setup guides in our instrument compatibility portal (https://www.thermofisher.com/instrumentsetup). If your instrument is not listed there, please contact Drug Discovery Technical Support at drugdiscoverytech@thermofisher.com.

Note: Test your microplate reader's TR-FRET setup before you begin any work with your assay. Using reagents that you have already purchased for your assay, you can test the microplate reader's TR-FRET setup. Please refer to the Terbium (Tb) Assay (http://tools.thermofisher.com/content/sfs/manuals/LanthaScreen_Tb_Instrument_Control_Application_Note17Sep10.pdf) and Europium (Eu) Assay (http://tools.thermofisher.com/content/sfs/manuals/Eu_Instrument_Control_AppNote_12Nov10.pdf) Application Notes for plate reader setup.

For Research Use Only. Not for use in diagnostic procedures.