missing translation for 'onlineSavingsMsg'
Learn More

TPPS Hydrate (=Tetraphenylporphyrin Tetrasulfonic Acid Hydrate) 85.0+%, TCI America™

Catalog No. A5013100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5013100MG 100 mg
A50131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5013100MG Supplier TCI America Supplier No. A5013100MG

[Ultra-high sensitive spectrophotometric reagent for transition metals]

Chemical Identifiers

CAS 35218-75-8
Molecular Formula C44H30N4O12S4
Molecular Weight (g/mol) 934.98
MDL Number MFCD00070632
InChI Key YAVMDSYMZGJNES-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(4-sulfophenyl)porphyrin, Tetraphenylporphyrin Tetrasulfonic Acid
PubChem CID 49561
IUPAC Name 4-[10,15,20-tris(4-sulfophenyl)-21,24-dihydroporphyrin-5-yl]benzenesulfonic acid
SMILES C1=CC(=CC=C1C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)S(=O)(=O)O)C8=CC=C(C=C8)S(=O)(=O)O)C9=CC=C(C=C9)S(=O)(=O)O)N3)S(=O)(=O)O

Specifications

Color Blue
Quantity 100 mg
Formula Weight 934.98
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material TPPS Hydrate (=Tetraphenylporphyrin Tetrasulfonic Acid Hydrate) [Ultra-high sensitive spectro
RTECSNumber DB7339000
TSCA No