Learn More
Heathrow Scientific Tourniquets
Catalog No. 361069767
Change view
Click to view available options
Color:
Orange
Packaging Type:
Bulk Pack
Catalog No. Color Packaging Type
361069767 Orange Bulk Pack
1 options
Catalog No. 361069767 Supplier Heathrow Scientific Supplier No. HS120110
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Hi-viz orange alert for safer practice
  • Made from supple and smooth polyisoprene
  • Latex free
  • Less abrasive than other brands
  • No rubber proteins, resins or fatty acids that can produce hypersensitive reactions
  • Available in bulk or on a perforated roll
  • Uniform thickness prevents shredding and breaking
Specifications
Color Orange
Latex Free Yes
Material Polyisoprene
Quantity 250/Bulk
Product Type Tourniquet
Dimensions (L x W) 18 x 1 in. (45.7 x 2.5 cm)
Packaging Type Bulk Pack