Learn More
Brady™ ToughStripe Max Floor Marking Footprint
Catalog No. 17987990
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17987990 Red
17987985 Yellow
17987986 Green
17987987 White
17987988 Blue
17987989 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17987990 Supplier Brady™ Supplier No. 177149
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Built stronger to last longer on harsh industrial floors, ToughStripe Max Floor Marking Footprint is ideal for forklift and heavy traffic areas.
  • ToughStripe Max Floor Marking Footprint is purpose-built to hold its ground in forklift and heavy traffic areas.
  • 50% thinner than previous version of ToughStripe Max and twice as tough as similar-thickness vinyl tape. T
  • Thinner profile and true edge-to-edge adhesive minimize pick points. Plasticizer-free polyester (PETG) material is more rigid than vinyl.
  • Every layer is hand selected, pressure tested and purpose built to protect colors and outlast the competition.
  • Damage-isolating design keeps tape intact longer by preventing rips, gouges and tears from running.
  • Matte finish provides excellent color contrast for clear visibility under any lighting.
Specifications
Product Type Floor Marking Tape
Material Polyester
Dimensions (L x W) 4 x 10 in.
Color Red
Adhesive Type Rubber