Learn More
Brady™ ToughStripe™ Die-Cut Floor Marking Letter X
ToughStripe™ Die-Cut Floor Marking (Letter X) is used for marking aisle-ways, shipping and receiving docks, distribution, or other areas of your facility.
Supplier: Brady™ 121831
Description
- Visible floor-marking improves safety and organization in almost any industrial environment
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Tape removes cleanly without any unsightly adhesive residue or damage to the floor
- Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.22
Chemical Identifiers
| Permanent Rubber-based |
Specifications
| Permanent Rubber-based | |
| Red |