Learn More
Brady™ ToughStripe™ Die-Cut Floor Marking Letter W
Catalog No. 19108277
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19108277 Black
19108278 White
19108279 Red
19108280 Green
19108281 Blue
19108282 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19108277 Supplier Brady™ Supplier No. 121823
ToughStripe™ Die-Cut Floor Marking (Letter W) is used for marking aisle-ways, shipping and receiving docks, distribution, or other areas of your facility.
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Tape removes cleanly without any unsightly adhesive residue or damage to the floor
  • Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.22
Specifications
Adhesive Type Permanent Rubber-based
Color Black