missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Cold Floor Marking Tape, Polyester

Catalog No. 17111337
Change view
Click to view available options
Color:
Black
Black, White
Black, Yellow
Blue
Green
Orange
Red
White
Yellow
Dimensions (L x W):
100 ft. x 2 in. (30.48 m x 5.08 cm)
100 ft. x 3 in. (30.48 m x 7.62 cm)
100 ft. x 4 in. (30.48 m x 10.16 cm)
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
17111337 Black, Yellow 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111314 White 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111315 White 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111316 White 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111317 Black 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111318 Black 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111319 Black 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111320 Blue 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111321 Blue 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111322 Blue 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111323 Green 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111324 Green 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111325 Green 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111326 Orange 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111327 Orange 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111328 Orange 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111329 Red 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111330 Red 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111331 Red 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111332 Black, White 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111333 Black, White 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111334 Black, White 100 ft. x 4 in. (30.48 m x 10.16 cm)
17111335 Black, Yellow 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111336 Black, Yellow 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111311 Yellow 100 ft. x 2 in. (30.48 m x 5.08 cm)
17111312 Yellow 100 ft. x 3 in. (30.48 m x 7.62 cm)
17111313 Yellow 100 ft. x 4 in. (30.48 m x 10.16 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Catalog No. 17111337 Supplier Brady™ Supplier No. 152756

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use this durable floor tape to mark refrigerators, freezers and other cold surfaces. Ideal for food and beverage refrigerated warehousing and other cold storage facilities.

  • Can be applied to cold surfaces as low as -13°F (-25°C) for reliable placement in extreme cold.
  • Aggressive adhesive performance ensures long-lasting hold in demanding environments.
  • Easy installation and removal supports efficient facility layout updates.
  • Withstands heavy foot traffic and medium vehicle traffic for durable use in high-activity areas.
  • Suitable for organization within large cold storage areas and improves visual management.
  • Can be applied using the Area Marking Tape Applicator (part number 804314) for faster installation.

Specifications

Product Type Floor Marking Tape
Material Polyester
Dimensions (L x W) 100 ft. x 4 in. (30.48 m x 10.16 cm)
Adhesive Type Permanent Acrylic
Color Black, Yellow