Research Products Corporation Tough-Spots for Laser Printers, 1/2 Inch Diameter, Yellow, 3,840 per Package
Catalog No. 50212849
Tough-Spots are chemically resistant labels that adhere to polypropylene tubes without cracking, peeling or degrading. Specially designed to withstand boiling water baths, autoclaving freezing, cryogenic liquid nitrogen storage, organic solvents and caustic agents. Tough-Spots are made of a special smear resistant material and will accept any marking instrument.
Catalog No.
50212849
Supplier
Research Products CorporationSupplier No.
247139Y
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tough-Spots are chemically resistant labels that adhere to polypropylene tubes without cracking, peeling or degrading. Specially designed to withstand boiling water baths, autoclaving freezing, cryogenic liquid nitrogen storage, organic solvents and caustic agents. Tough-Spots are made of a special smear resistant material and will accept any marking instrument.