UserName
Tough-Spots, large violet labels for 1.5 ml and 2.0 ml tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.