missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems TotalStain Q, PVDF Membranes

Catalog No. AC2225AZ
Change view
Click to view available options
Detection Method:
Fluorescense
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
AC2225AZ Fluorescense
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC2225AZ Supplier Azure Biosystems Supplier No. AC2225

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescent total protein stain for PVDF membranes. Sufficient amount for 15 mini-blots. 600 mL kit

  • Fluorescent Total Protein Stain for Western blots
  • Accurate and quantitative fluorescent total protein stain to perform normalization of Western blots. Blots can be imaged on both camera-based and scanner instruments with green channel or Safe-Dyes channel.
  • TotalStain Q only adds a short 24-minute step to your Western blot protocol, between protein transfer and blocking buffer incubation. It is compatible with subsequent Western blot detection using chemiluminescence or Near Infrared (NIR) fluorescence.
  • Kit contains enough material for 15 membranes

Specifications

Color Clear
Concentration 1X
Detection Method Fluorescense
Format Bottle
Formulation Liquid
Storage Requirements Room Temperature
Product Type Protein Stain