missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Total Cholesterol Reagents

Catalog No. TR13421
Change view
Click to view available options
Quantity:
2 x 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR13421 2 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR13421 Supplier Thermo Scientific™ Supplier No. JTR13421
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Determine quantity of cholesterol in serum in vitro with Thermo Scientific™ Total Cholesterol Reagents.

Measurement of serum cholesterol levels can serve as an indicator of liver, biliary or thyroid function, intestinal absorption, adrenal disease and predisposition toward coronary artery disease. Increased levels are associated with nephrosis, diabetes mellitus and obstructive jaundice. When used in conjunction with triglyceride, HDL and LDL determinations, total cholesterol assays provide valuable information for the prediction of coronary artery disease. Decreased levels may be observed in cases of hyperthyroidism, certain anemias, malabsorption and wasting syndromes.
  • Up to 12 months reconstituted stability and various kit sizes to minimize reagent wastage
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers available on request

Specifications

Certifications/Compliance CE Marked
Description Infinity Cholesterol 2 x 125mL
CE Marker Yes
Detectable Analytes Cholesterol
Sample Type Plasma, Serum, Serum or Plasma
Type Reagent
Linearity Up to 20mmol/L (774mg/dL)
Methods Compatibility Endpoint
Quantity 2 x 125 mL
Stability Until expiry at 2-8°C