Learn More
Fujifilm Healthcare Americas Corporation Cholesterol E
Supplier:  Fujifilm Healthcare Americas Corporation 99902601
Encompass
The Cholesterol E kit is an enzymatic colorimetric method for the quantitative determination of total cholesterol in serum. Package size: Kit includes: Buffer Solution- 2 x 150 mL, Color Reagent- 2 x for 150 mL, Standard Solution- 1 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9138103
Only null left