missing translation for 'onlineSavingsMsg'
Learn More

Diazyme Laboratories Total Bile Acids Control

Catalog No. 23700586
Encompass
Change view
Click to view available options
Format: 2 x 1 mL vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23700586 Format: 2 x 1 mL vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23700586 Supplier Diazyme Laboratories Supplier No. DZ042ACON

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diazyme Total Bile Acids Control Set is used for the testing of Total Bile Acids in Serum, EDTA Plasma, and Lithium Heparin Plasma.

  • A liquid stable system, ready to use for both manual methods and is adaptable for many automated chemistry analyzers
  • The assay has excellent sensitivity with a linear range from 0 - 180 μM
  • Unique enzyme cycling method amplifies signal for accurate and fast measurement using a small sample up to 50% less than conventional NBT methods making it an excellent choice
  • The assay has excellent precision with Intra-Assay Precision CV% of <4% and Inter-Assay Precision CV% of <3%
  • The following substances normally present in serum produced less than 10% deviation at the listed concentrations: Triglycerides at 750 mg/dL, Ascorbic acid at 50 mg/dL, Bilirubin at 50 mg/dL and Hemoglobin at 500 mg/dL
  • Packaged for optimal operator convenience reagent transfer can be eliminated for most chemistry systems with instrument specific packaging options including RocheTM Hitachi 917 series, Beckman AU (400/600/640/680), Beckman Synchron CX, and DXC

Specifications

Certifications/Compliance 510(k) Cleared, EU: CE IVD, Health Canada Registered
Description Total Bile Acids (Enzyme Cycling) Assay Control Set
Concentration Lab dependent
Format 2 x 1 mL Vial
Type Diagnostic