missing translation for 'onlineSavingsMsg'
Learn More

Diazyme Laboratories Total Bile Acids Calibrator

Catalog No. 23700584
Encompass
Change view
Click to view available options
Concentration:
Lab dependent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23700584 Lab dependent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23700584 Supplier Diazyme Laboratories Supplier No. DZ042ACAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diazyme Total Bile Acids Calibrator Set is used for the testing of Total Bile Acids in Serum, EDTA Plasma, and Lithium Heparin Plasma.

  • A liquid stable system, ready to use for both manual methods and is adaptable for many automated chemistry analyzers
  • The assay has excellent sensitivity with a linear range from 0 - 180 μM
  • Unique enzyme cycling method amplifies signal for accurate and fast measurement using a small sample up to 50% less than conventional NBT methods making it an excellent choice
  • The assay has excellent precision with Intra-Assay Precision CV% of <4% and Inter-Assay Precision CV% of <3%
  • The following substances normally present in serum produced less than 10% deviation at the listed concentrations: Triglycerides at 750 mg/dL, Ascorbic acid at 50 mg/dL, Bilirubin at 50 mg/dL and Hemoglobin at 500 mg/dL
  • Packaged for optimal operator convenience reagent transfer can be eliminated for most chemistry systems with instrument specific packaging options including RocheTM Hitachi 917 series, Beckman AU (400/600/640/680), Beckman Synchron CX, and DXC

Specifications

Certifications/Compliance 510(k) Cleared, EU: CE IVD, Health Canada Registered
Content And Storage Store at 2°C to 8°C; Note: Store CALIBRATOR tightly capped when not in use
Description Total Bile Acids (Enzyme Cycling) Assay Calibrator Set
Concentration Lab dependent
Format 1 x 2 mL Vial
Type Diagnostic
Includes One vial of liquid
Stability The total bile acid calibrator is stable until expiration date printed on label when stored properly at 2 to 8°C