missing translation for 'onlineSavingsMsg'
Learn More

Topiramate 98.0+%, TCI America™

Catalog No. T27555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27555G 5 g
T27551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27555G Supplier TCI America Supplier No. T27555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97240-79-4
Molecular Formula C12H21NO8S
Molecular Weight (g/mol) 339.359
MDL Number MFCD00865320
InChI Key KJADKKWYZYXHBB-XBWDGYHZSA-N
Synonym topiramate, topamax, epitomax, topamax sprinkle, tipiramate, topiramatum, tipiramato, topiramato, tipiramate french, tipiramato spanish
PubChem CID 5284627
ChEBI CHEBI:63631
IUPAC Name [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)COS(=O)(=O)N)C

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 339.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Topiramate
RTECSNumber LS7083000
TSCA No
Recommended Storage Refrigerator