missing translation for 'onlineSavingsMsg'
Learn More

Topiramate, 98%

Catalog No. AC456190050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456190050 5 g
AC456190010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456190050 Supplier Thermo Scientific Chemicals Supplier No. 456190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97240-79-4 | C12H21NO8S | 339.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97240-79-4
Molecular Formula C12H21NO8S
Molecular Weight (g/mol) 339.36
InChI Key KJADKKWYZYXHBB-XBWDGYHZSA-N
Synonym topiramate, topamax, epitomax, topamax sprinkle, tipiramate, topiramatum, tipiramato, topiramato, tipiramate french, tipiramato spanish
PubChem CID 5284627
ChEBI CHEBI:63631
IUPAC Name [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)COS(=O)(=O)N)C

Specifications

Melting Point 125.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Quantity 5 g
Formula Weight 339.36
Percent Purity 98%
Chemical Name or Material Topiramate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only