missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Top Grain Pigskin Leather Mig Tig Welding Gloves

Catalog No. 19065556A
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19065556A Small
19065556C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19065556A Supplier Fisherbrand™ Supplier No. 75320/S

Gloves are sewn with Aramid Fiber to provide strength and burn resistance.

  • Top Grain Pigskin leather is smooth and breathable due its porous texture
  • Leather becomes softer with use and withstands moisture without stiffening
  • Stitched with Aramid Fiber for additional strength and burn resistance
  • Split leather gauntlet cuff provides additional forearm protection and allows for easy on/off
  • Straight thumb increases comfort in closed-fisted work
  • Unlined palm for dexterity

Specifications

Cuff Material Split Leather
Color Beige
Material Top Grain Pigskin Leather
Size Small
For Use With (Application) Automotive Assembly, MIG/TIG Welding
Glove Cuff Color Gray
Glove Cuff Style Gauntlet
Series 75-320
Length (English) 11 in.
Length (Metric) 28 cm