missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Top Dog 6 mil Die Cut 45 in. Apron

Top Dog 6 mil Die Cut 45 in. Apron is reversable and designed to fit comfortably over clothes.

Supplier:  Vikan™ 80016

 View more versions of this product

Catalog No. 17-227-941


Only null left
Explore available promotions

Description

Description

  • Reusable
  • Color-coded
  • FDA-compliant garments
  • Designed to support zoned color-coding programs
  • One-piece construction limits the potential for foreign body contamination and reduces harborage points for pathogens
  • Resistant to common sanitation chemicals, and impervious to animal fats and degreasers
Specifications

Specifications

Apron
Polyurethane
Reusable
Non-sterile
6 mil
45 x 45 in.
0.4 lb.
Universal
Yellow
Waist Ties
Ether-Based Polyurethane
FDA Compliant
45 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions