missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toluidine Blue O, Certified, 65% min dye content

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol

Supplier:  Thermo Scientific Chemicals J6601506

 View more versions of this product

Catalog No. AAJ6601506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Toluidine Blue O
C15H16ClN3S
Basic Blue 17,C.I. 52040
HNONEKILPDHFOL-UHFFFAOYSA-M
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
7083
≥65% dye content
5 g
92-31-9
MFCD00011934
Soluble in water at 1mg/ml
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
305.82
305.83
Glass bottle
Powder

Chemical Identifiers

92-31-9
305.82
HNONEKILPDHFOL-UHFFFAOYSA-M
7083
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
C15H16ClN3S
MFCD00011934
Basic Blue 17,C.I. 52040
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-146-2

RTECSNumber : SP5670000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only