missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl chloride, 98%

Catalog No. AC170490050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170490050 5 g Glass bottle
AC170490250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170490050 Supplier Thermo Scientific Chemicals Supplier No. 170490050

CAS: 1939-99-7 | C7H7ClO2S | 190.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1939-99-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007455
InChI Key OAHKWDDSKCRNFE-UHFFFAOYSA-N
Synonym alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
PubChem CID 74740
IUPAC Name phenylmethanesulfonyl chloride
SMILES C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 94°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.6240 to 1.626
Linear Formula C6H5CH2SO2Cl
Quantity 5 g
Beilstein 11, 116
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in ethanol and ether
Formula Weight 190.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Toluenesulfonyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 217-717-1
TSCA TSCA

RUO – Research Use Only