Learn More
Thermo Scientific™ Toluene in Hexane Solution Standard
Catalog No. 14386450
Change view
Click to view available options
For Use With (Equipment):
Evolution 60S, 201, 220 260 Bio, 300, GENESYS 10S, BioMate 3S
Catalog No. For Use With (Equipment)
14386450 Evolution 60S, 201, 220 260 Bio, 300, GENESYS 10S, BioMate 3S
1 options
Catalog No. 14386450 Supplier Thermo Scientific™ Supplier No. 222226600
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ traceable Toluene in Hexane Solution Standard ensures accurate wavelength and absorbance results by verifying spectrophotometer resolution.
Simple checks of the resolution and bandwidth of your spectrophotometer will ensure that the best possible results are obtained for your analyses. A 0.02% Toluene in Hexane Solution is the pharmacopoeia preferred method for verifying resolution of a spectrophotometer.
  • Heat-sealed in a quartz cuvette and traceable to the NIST™
  • Resolution is measured by calculating the ratio of the peak maixma of toluene at 269nm to its peak minima at 266nm
  • Resulting ratio provides a measure of instrument resolution
  • To determine spectral bandwidth, compare ratio obtained with table provided with the standard
Specifications
Description Resolution Standard; Verification Standard
Type Toluene in Hexane Solution Standard
Wavelength Range 265 to 270 nm
For Use With (Equipment) Evolution 60S, 201, 220 260 Bio, 300, GENESYS 10S, BioMate 3S