missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS Reagent, Reagents

Catalog No. 01337647
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Glass Bottle
Safety Coated Glass
Steel Drum
Steel Pail
Tin Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337647 200 L Steel Drum
01337646 20 L Steel Pail
01337645 4 L Glass Bottle
01337644 4 L Safety Coated Glass
01337643 4 L Tin Can
01337642 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337647 Supplier Labchem Inc Supplier No. C1341000200E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Toluene, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Toluene, ACS Reagent
Melting Point -95°C
CAS 108-88-3
Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.866
Molecular Formula C6H5CH3
Synonym Methylbenzene, Phenylmethane, Toluol
Boiling Point 111°C
Molecular Weight (g/mol) 92.1384
Grade ACS
Show More Show Less