missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolnaftate
Description
Tolnaftate is used to study the mechanism of ergosterol (sterol) biosynthesis at the rate-limiting step of squalene epoxidase inhibition. Tolnaftate is clinically used to treat cutaneous infections such as athlete?s foot, jock itch, and ringworm1. It also has been used as a comparator compound for antifungal efficiency studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Specifications
Specifications
| Type | Tolnaftate |
| Physical Form | Powder |
| Melting Point | 110°C to 113°C |
| Color | White |
| Quantity | 5 g |
| Merck Index | 14,9519 |
| Solubility Information | Soluble in chloroform at 50 mg/ml; Sparingly soluble in ethanol or methanol |
| Formula Weight | 307.41 |
| Chemical Name or Material | Tolnaftate |
Safety and Handling
| EINECSNumber | 219-266-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only