missing translation for 'onlineSavingsMsg'
Learn More

Tolnaftate 97.0+%, TCI America™

Catalog No. T30591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30591G 1 g
T30595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30591G Supplier TCI America Supplier No. T30591G

Chemical Identifiers

CAS 2398-96-1
Molecular Formula C19H17NOS
Molecular Weight (g/mol) 307.411
MDL Number MFCD00056611
InChI Key FUSNMLFNXJSCDI-UHFFFAOYSA-N
Synonym tolnaftate, tolnaphthate, tinactin, tonoftal, sporiline, tinaderm, tolsanil, aftate, chinofungin, dermoxin
PubChem CID 5510
ChEBI CHEBI:9620
IUPAC Name O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
SMILES CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 113°C
Color White
Quantity 1 g
Formula Weight 307.41
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tolnaftate
EINECSNumber (4)-1110
RTECSNumber FD8891300
TSCA Yes
Recommended Storage Refrigerator