missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tolazoline hydrochloride, 99%

Catalog No. p-3758818
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138980500 50 g Plastic bottle
AC138982500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138980500 Supplier Thermo Scientific Chemicals Supplier No. 138980500

CAS: 59-97-2 | C10H12N2·HCl | 196.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-97-2
Molecular Formula C10H12N2·HCl
Molecular Weight (g/mol) 196.72
MDL Number MFCD00012693
InChI Key RHTNTTODYGNRSP-UHFFFAOYSA-N
Synonym tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride
PubChem CID 6048
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC=C2.Cl

Specifications

Melting Point 172.0°C to 176.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 9666
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in chloroform, freely soluble in alcohol, slightly soluble in ether and ethyl acetate
Formula Weight 196.72
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tolazoline hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-447-3

RUO – Research Use Only