Learn More
Veolia Wts Analytical System Suitability Set - 800/900 Series, Veolia DFS Item
Supplier:  GE Analytical Instruments STD3100401
System suitability set includes 1 Rw - reagent water control, 1 Rs - standard solution and 1 Rss - system suitability solution.
Catalog No. NC9285532
Only null left
Explore available promotions