Learn More
Thermo Scientific™ TOC Certified Containers
GREENER_CHOICE
Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.
Supplier:  Thermo Scientific™ 24 414/TS/WS 2
 View more versions of this product
Catalog No. 12200164
Only null left
Description
Description
Trust our low-level certified vials for Total Organic Carbon testing and sampling. Manufactured, tested and packaged by exacting standards,Thermo Scientific™ TOC Certified Containers are certified to contribute <10ppb or < 20ppb TOC as background.
Containers:
  • Available in several sizes and styles of containers, including the popular 40mL autosampler vials cleaned, certified and ready to use
  • Include Certificate of Analysis
Includes:
  • Sampling and storage containers
  • Autosampler compatible vials
  • Culture tubes
  • Low TOC water
Recommended for:
  • TOC testing
  • USP method 643 testing
  • Off-line grab sampling of high purity water
  • Storage of validation standards.
Specifications
Specifications
24-414 Polypropylene Cap with Septa, Pre-Cleaned
Polypropylene
72
Polypropylene
72 Cs.
Certified to contribute <10ppb, <20ppb, Certificate of Analysis (COA), Certificate of Conformance (COC)
PTFE/Silicone
Pre-cleaned
Cap with Septa
Screw Cap with Septum
24-414
Polypropylene Cap
Product Suggestions
Product Suggestions