missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin 94.0+%, TCI America™

Catalog No. T25035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25035G 5 g
T250325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25035G Supplier TCI America Supplier No. T25035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32986-56-4 | Formula: C18H37N5O9 | Weight: 467.52 g/mol

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
MDL Number MFCD00077885
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym O-[3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)]-O-[2,6-diamino-2,3,6-trideoxy-alpha-D-ribo-hexopyranosyl-(1->4)]-2-deoxy-D-streptamine
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Type Tobramycin
Physical Form Crystalline Powder
Melting Point 178°C
Color White/Yellow
Quantity 5 g
Formula Weight 467.52
Percent Purity ≥94.0% (T)
Grade Not Available
Chemical Name or Material Tobramycin
RTECSNumber WK2100000
TSCA No