missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tobramycin 94.0+%, TCI America™
Supplier: TCI America T250325G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 32986-56-4 | |
| 467.52 | |
| NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
| 36294 | |
| (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol |
| C18H37N5O9 | |
| MFCD00077885 | |
| O-[3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)]-O-[2,6-diamino-2,3,6-trideoxy-alpha-D-ribo-hexopyranosyl-(1->4)]-2-deoxy-D-streptamine | |
| CHEBI:28864 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N |
Specifications
| Tobramycin | |
| Crystalline Powder | |
| 178°C | |
| 25 g | |
| MFCD00077885 | |
| NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
| (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol | |
| 36294 | |
| 467.52 |
| Tobramycin | |
| 32986-56-4 | |
| White/Yellow | |
| C18H37N5O9 | |
| O-[3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)]-O-[2,6-diamino-2,3,6-trideoxy-alpha-D-ribo-hexopyranosyl-(1->4)]-2-deoxy-D-streptamine | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N | |
| 467.52 | |
| CHEBI:28864 | |
| ≥94.0% (T) |
Safety and Handling
RTECSNumber : WK2100000
TSCA : No