missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin, MP Biomedicals™

Catalog No. p-3593055
Change view
Click to view available options
Quantity:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15692625 25 mg
ICN15692690 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15692625 Supplier MP Biomedicals Inc Supplier No. 0215692625

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym Distobram, Gernebcin
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Type Tobramycin
Physical Form Powder
Color White
Quantity 25 mg
Formula Weight 467.5
Chemical Name or Material Tobramycin