missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin, 900μg/mg

Catalog No. AAJ6604003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6604003 1 g
AAJ6604006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6604003 Supplier Thermo Scientific Chemicals Supplier No. J6604003

CAS: 32986-56-4 | C18H37N5O9 | 467.52 g/mol

Primarily targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
MDL Number MFCD00077885
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym tobramycin, nebramycin 6, nebramycin factor 6, nebramycin vi, tobramicin, tobramycetin, nebcin, 3'-deoxykanamycin b, aktob, tobi
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Packaging Plastic bottle
Quantity 1 g
Beilstein 1357507
Solubility Information Soluble in water
Formula Weight 467.51
Physical Form Powder
Chemical Name or Material Tobramycin
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-322-5
RTECSNumber WK2100000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only