missing translation for 'onlineSavingsMsg'
Learn More

Promega TNT™ Coupled Wheat Germ Extract Systems

Catalog No. PRL4130
Encompass
Change view
Click to view available options
Product Type:
TNT SP6 Coupled Wheat Germ Extract System
TNT T7 Coupled Wheat Germ Extract System
TNT T7/SP6 Coupled Wheat Germ Extract System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRL4130 TNT SP6 Coupled Wheat Germ Extract System
PRL5030 TNT T7/SP6 Coupled Wheat Germ Extract System
PRL4140 TNT T7 Coupled Wheat Germ Extract System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRL4130 Supplier Promega Supplier No. L4130
Only null left

Includes

TNT Wheat Germ Extract, Appropriate TNT wheat germ polymerase, Control DNA, TNT Reaction Buffer, Luciferase assay reagent, Luciferase assay wells, Amino acid mixtures

The TNT Coupled Wheat Germ Extract Systems are one-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.

  • Single-Tube Coupled Transcription/Translation System
  • Produces 2- to 6-fold more protein than standard wheat germ systems
  • Produce protein from linearized DNA in 1.5 hours
  • You choose the promoter

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patents 649289.
  • U.S. Patents 5,324,637 and 5,492,817; Australian Patent 660329.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.

Specifications

Content And Storage <-65°C
For Use With (Application) Translation of mammalian transcription factors
Includes 1mL TNT™ Wheat Germ Extract, 90μL TNT™ Reaction Buffer, 60μL TNT™ RNA Polymerase (SP6, T3 or T7, individually or 2 x 30μL when two Polymerases are
No. of Reactions 40
Product Type TNT SP6 Coupled Wheat Germ Extract System