missing translation for 'onlineSavingsMsg'
Learn More

TMS-Imidazole (=N-Trimethylsilylimidazole) 98.0+%, TCI America™

Catalog No. A56055ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56055ML Supplier TCI America Supplier No. A56055ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Melting Point -42°C
Color Colorless
Boiling Point 94°C
Quantity 5 mL
UN Number 1993
Formula Weight 140.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material TMS-Imidazole (=N-Trimethylsilylimidazole) [for Gas Chromatography]
RTECSNumber NI8700000
TSCA Yes
Recommended Storage Refrigerator