missing translation for 'onlineSavingsMsg'
Learn More

TMS-HT (=HMDS and TMCS in Anhydrous Pyridine) [Trimethylsilylating Reagent, for OH compounds], TCI America™

Catalog No. t027412ml
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T027412ML 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T027412ML Supplier TCI America Supplier No. T027412ML

Chemical Identifiers

MDL Number MFCD00192427

Specifications

Color White-Yellow
Quantity 12 mL
Physical Form Clear Liquid-Cloudy Liquid at 20°C
UN Number 2924
Chemical Name or Material TMS-HT (=HMDS and TMCS in Anhydrous Pyridine) [Trimethylsilylating Reagent, for OH compounds]
TSCA No
Recommended Storage Refrigerator