missing translation for 'onlineSavingsMsg'
Learn More

TMS-BA [=N,O-Bis(trimethylsilyl)acetamide] (25% in Acetonitrile) [for Gas Chromatography], TCI America™

Catalog No. A56025ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56025ML Supplier TCI America Supplier No. A56025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Color Yellow
Quantity 5 mL
UN Number 1992
Physical Form Liquid
Chemical Name or Material TMS-BA [=N,O-Bis(trimethylsilyl)acetamide] (25% in Acetonitrile) [for Gas Chromatography]
EINECSNumber (2)-3273
RTECSNumber AK3000000
TSCA Yes
Recommended Storage Refrigerator