missing translation for 'onlineSavingsMsg'
Learn More

TMPyP 98.0+%, TCI America™

Catalog No. A5014100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5014100MG 100 mg
A50141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5014100MG Supplier TCI America Supplier No. A5014100MG

[Ultra-high sensitive spectrophotometric reagent for Cu, Mg], [For the simultaneous determination of metals by HPLC]

Chemical Identifiers

CAS 36951-72-1
Molecular Formula C72H66N8O12S4
Molecular Weight (g/mol) 1363.604
MDL Number MFCD00013468
InChI Key AKZFRMNXBLFDNN-UHFFFAOYSA-K
Synonym tmpyp, tmpyp4 tosylate, tmpyp tetratosylate, tmpyp4 tetratosylate, tppt 4, meso-tetra 4-n-methyl-pyridyl porphyrin, meso-tetrakis n-methyl-4-pyridyl porphine tetratosylate salt, meso-tetrakis n-methylpyridinium-4-yl porphine tetratosylate, meso-tetrakis n-methyl-4-pyridyl porphine tetrakis p-toluenesulfonate, 5,10,15,20-tetrakis 1-methylpyridinium-4-yl porphyrin tetra p-toluenesulfonate
PubChem CID 11979833
IUPAC Name 4-methylbenzenesulfonate;5,10,15,20-tetrakis(1-methylpyridin-1-ium-4-yl)-21,22-dihydroporphyrin
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=[N+](C=C7)C)C8=CC=[N+](C=C8)C)C9=CC=[N+](C=C9)C)N3

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 1363.60
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material TMPyP [=alpha,beta,gamma,delta-Tetrakis(1-methylpyridinium-4-yl)porphyrin p-Toluenesulfonate]