missing translation for 'onlineSavingsMsg'
Learn More

Monoethanolamine, NF, 98-100.5%, Spectrum™ Chemical

Catalog No. 18604679
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604679 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604679 Supplier Spectrum Chemical Mfg Cor Supplier No. M1322500ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1322, 141-43-5, C2H7NO

Spectrum™ Chemical Monoethanolamine, NF - All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

CAS Max % 1
Boiling Point 170.8°C
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 98 to 100.5%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Monoethanolamine