missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TLR7 Antibody Blocking Peptide

Catalog No. NB398691
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398691 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398691 Supplier R&D Systems™ Supplier No. NB10056588PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Gene ID (Entrez) 51284
Product Line Novus
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56588
Gene Alias toll-like receptor 7
Gene Symbol TLR7
Quantity 0.1 mg
Research Category Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Storage Buffer Peptide dissolved in dH20. Contains no BSA.