missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ TLC Silica Gel 60 Plate with F254 Coating

Catalog No. M1055700001
Change view
Click to view available options
Quantity:
25 Units/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1055700001 25 Units/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1055700001 Supplier MilliporeSigma™ Supplier No. 1055700001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for thin layer chromatography applications

  • Combination of silica gel 60, addition of polymeric binder
  • Adherent hard surface that will not crack or blister
  • Size: 10 x 20cm

Specifications

Backing Material Aluminum Support
Color White
Quantity 25 Units/Pk.
Type Silica Gel
For Use With (Application) Thin Layer Chromatography
Storage Requirements Room temperature