missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ TLC Saturation Pads

Catalog No. 111011158
Change view
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
111011158 10 x 20 cm
111016153 20 x 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111011158 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2652331PAK

Pads improve reproducibility and reduce 'edge effects' when performing thin layer chromatography.

Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor.

Specifications

Quantity 100/Pk.
For Use With (Application) TLC
Packing Material Pk. of 100 Ea.
Dimensions (L x W) 10 x 20 cm
Length (Metric) 10 cm
Width (Metric) 20 cm