Learn More
MilliporeSigma™ Supelco™ TLC Plate Storage Racks
Used to carry and store TLC plates without damage to media.
$714.01
Specifications
Material Stainless Steel (20 Gauge)
For Use With (Application) TLC
View More Specs
Products 1
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
111009367
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z2660941EA
20 x 20 cm Plates
Each for $714.01
Only null left
Description
Description
Racked plates may be air or oven dried. Holds up to 10 standard plates in either a vertical or horizontal position. Racks are stainless steel with a carrying handle.
Specifications
Specifications
Stainless Steel (20 Gauge)
TLC