missing translation for 'onlineSavingsMsg'
Learn More

Titriplex III, For Analysis, (Ethylenedinitrilotetraacetic Acid, Disodium Salt Dihydrate), ACS, MilliporeSigma™

Catalog No. p-7082932
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Fiber Carton
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1084180100 100 g Fiber Carton
M1084180250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1084180100 Supplier Technidata Supplier No. 1.08418.0100
Only null left

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 110°C (Elimination of water of crystallisation)
pH 4 to 5 (50g/L, H2O, 20°C)
Packaging Fiber Carton
Quantity 100 g
Solubility Information 100g/L
Formula Weight 372.24
EINECSNumber 205-358-3
RTECSNumber AH4410000