missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titriplex III, For Analysis, (Ethylenedinitrilotetraacetic Acid, Disodium Salt Dihydrate), ACS, MilliporeSigma™
Specifications
Specifications
| Melting Point | 110°C (Elimination of water of crystallisation) |
| pH | 4 to 5 (50g/L, H2O, 20°C) |
| Packaging | Fiber Carton |
| Quantity | 100 g |
| Solubility Information | 100g/L |
| Formula Weight | 372.24 |
| Grade | ACS |
Safety and Handling
| EINECSNumber | 205-358-3 |
| RTECSNumber | AH4410000 |