Learn More
Eisco™ Titration Labware Set
Catalog No. S02232
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
S02232 None
1 options
Catalog No. S02232 Supplier Eisco™ Supplier No. FSE202301
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
All materials required to perform titrations in one convenient package
  • Convenient set eliminates the need to piece together a laboratory titration setup
  • Includes six pieces of essential titration glassware, all made of heavy-duty borosilicate 3.3 glass: (1) Burette, 50 mL; (2) Beakers, 100 mL and 250 mL; (2) Graduated Cylinders, 50 mL and 100 mL; (1) Erlenmeyer Flask, 250 mL.
  • Also includes a five-piece, industrial-quality metal retort stand set: (1) Base made of corrosion resistant, powder coated steel, 8 x 5 in., 1.75 lbs.; (1) Rod made of stainless steel, 23.6 in. L x 10 mm dia. with 10 mm by 1.5 mm metric thread; (1) Support Ring; 2.5 in. dia.; (1) Four-pronged cork-lined Clamp; 10.5 cm max opening; (1) Bosshead.
  • All glassware items are packaged in a custom fit, thick foam casing, for safe storage and worry-free transport
  • All metal items come packed in a custom fit box for convenient storage when disassembled
Specifications
Burette Functions Manual Key Stopcock Control
Functions Available Manual Titration
Burette Sizes 50 mL
Calibration Calibrated to Contain
Display Type None
Type Titration Labware
For Use With (Application) Lab bench titrations
Includes (1) Burette, (2) Beakers, (2) Graduated Cylinders, (1) Erlenmeyer Flask, (1) Retort Base, (1) Support Rod, (1) Support Ring, (1) Clamp, (1) Bosshead.
Material Borosilicate Glass and Steel
Dimensions (L x W x H) 8 x 5 x 24 in.