missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Titertek™ Pipettor 12-channel, 50–300μL Adjustable Volume
Description
- Ejector works with dispensing head in any position
- 8-and 12-channel pipettors are available for use with standard 96-well plates
- Individual piston/tip cone assemblies allow easy repair and maintenance
- Compound material tip cone design allows visual seal verification
Specifications
Specifications
| For Use With | Standard 96-Well Plates |
| Number of Channels | 12 |
| Product Line | Titertek |
| Type | Pipette |
| Volume (Metric) | 50 to 300 μL |