missing translation for 'onlineSavingsMsg'
Learn More

Titanocene Dichloride 98.0+%, TCI America™

Catalog No. p-7136364
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T06165G 5 g
T061625G 25 g
2 options

Catalog No. T06165G

Supplier: TCI America T06165G

Only null left

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Bis(cyclopentadienyl)titanium Dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

Quantity 5 g
Formula Weight 248.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Titanocene Dichloride
EINECSNumber (3)-4343
RTECSNumber XR2050000
TSCA Yes