missing translation for 'onlineSavingsMsg'
Learn More

Titanium wire, 0.75mm (0.03in) dia, 99.98% (metals basis)

Catalog No. p-7048214
Change view
Click to view available options
Quantity:
25 cm
100 cm
Catalog No. Quantity
AA00361BU 25 cm
AA00361BY 100 cm
2 options

Catalog No. AA00361BU

Supplier: Thermo Scientific Chemicals 000361BU

Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 25 cm
Odor Odorless
Weight ≈1.99g/m
Form Wire
Percent Purity 99.98%
Assay (metals basis)
Chemical Name or Material Titanium wire, 0.75mm (0.03 in.) dia., Temper: as drawn
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only